Accessories for glassware machine washer
64, PAD2 booster pump for non pressurised RO or DI water feed, where tank is positioned at floor level. Requires PAD2R when installed in Basic Line models, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW2045, GW3060BX, GW3060BXC, GW3060P, GW3060SC, GW4060, GW4060C, GW4060S, GW4060SC, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Catalog # 141-0764
Supplier: SMEG
Jump to:
Product Details & Documents
Specifications
- Popis:PAD2 booster pump for non pressurised RO or DI water feed, where tank is positioned at floor level. Requires PAD2R when installed in Basic Line models
- Sekvence:64
- Určeno pro:GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW2045, GW3060BX, GW3060BXC, GW3060P, GW3060SC, GW4060, GW4060C, GW4060S, GW4060SC, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC