Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
Accessories for glassware machine washer
64, PAD2 booster pump for non pressurised RO or DI water feed, where tank is positioned at floor level. Requires PAD2R when installed in Basic Line models, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW2045, GW3060BX, GW3060BXC, GW3060P, GW3060SC, GW4060, GW4060C, GW4060S, GW4060SC, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Catalog # 141-0764
Supplier:   SMEG
Accessories for glassware machine washer
Catalog # 141-0764
Supplier:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
64, PAD2 booster pump for non pressurised RO or DI water feed, where tank is positioned at floor level. Requires PAD2R when installed in Basic Line models, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW2045, GW3060BX, GW3060BXC, GW3060P, GW3060SC, GW4060, GW4060C, GW4060S, GW4060SC, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Price
15 690,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    PAD2 booster pump for non pressurised RO or DI water feed, where tank is positioned at floor level. Requires PAD2R when installed in Basic Line models
  • Sekvence:
    64
  • Určeno pro:
    GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW2045, GW3060BX, GW3060BXC, GW3060P, GW3060SC, GW4060, GW4060C, GW4060S, GW4060SC, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.