Accessories for glassware machine washer
88, Universal flask washing, stainless steel, with 40 positions, for washing narrow necked glasswares, with drying system, GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C
GW4090C
Katalogové číslo 141-0736
Dodavatel: SMEG
Přejít na:
Product Details & Documents
Specifikace
- Popis:Universal flask washing, stainless steel, with 40 positions, for washing narrow necked glasswares, with drying system
- Sekvence:88
- Určeno pro:GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C