Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
88, Universal flask washing, stainless steel, with 40 positions, for washing narrow necked glasswares, with drying system, GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C
Katalogové číslo 141-0736
Dodavatel:   SMEG
Accessories for glassware machine washer
Katalogové číslo 141-0736
Dodavatel:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
88, Universal flask washing, stainless steel, with 40 positions, for washing narrow necked glasswares, with drying system, GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C
Přejít na:
Price
53 300,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Universal flask washing, stainless steel, with 40 positions, for washing narrow necked glasswares, with drying system
  • Sekvence:
    88
  • Určeno pro:
    GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
    GW4090C