Accessories for glassware machine washer
52, LPM2020DS jet rack, stainless steel, with drying system, with 20 pipette jets plus 20 assorted jet nozzles, GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Katalogové číslo 141-0746
Dodavatel: SMEG
Přejít na:
Product Details & Documents
Specifikace
- Popis:LPM2020DS jet rack, stainless steel, with drying system, with 20 pipette jets plus 20 assorted jet nozzles
- Sekvence:52
- Určeno pro:GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC