Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
52, LPM2020DS jet rack, stainless steel, with drying system, with 20 pipette jets plus 20 assorted jet nozzles, GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Katalogové číslo 141-0746
Dodavatel:   SMEG
Accessories for glassware machine washer
Katalogové číslo 141-0746
Dodavatel:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
52, LPM2020DS jet rack, stainless steel, with drying system, with 20 pipette jets plus 20 assorted jet nozzles, GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Přejít na:
Price
58 500,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    LPM2020DS jet rack, stainless steel, with drying system, with 20 pipette jets plus 20 assorted jet nozzles
  • Sekvence:
    52
  • Určeno pro:
    GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC