Accessories for glassware machine washer
6, Auxiliary tank (50 L) in PE-HD for demineralised water with automatic level control for external water feeding; compatible with demi water booster pump PAD/PAD2; water intake from auxiliary system
Dimensions (Ø×H): 340×640 mm, −
Dimensions (Ø×H): 340×640 mm, −
141-0793
: SMEG
- Popis:Auxiliary tank (50 L) in PE-HD for demineralised water with automatic level control for external water feeding; compatible with demi water booster pump PAD/PAD2; water intake from auxiliary system
Dimensions (Ø×H): 340×640 mm - Sekvence:6
- Určeno pro:−