Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
6, Auxiliary tank (50 L) in PE-HD for demineralised water with automatic level control for external water feeding; compatible with demi water booster pump PAD/PAD2; water intake from auxiliary system
Dimensions (Ø×H): 340×640 mm, −
  141-0793
 :   SMEG
Accessories for glassware machine washer
  141-0793
 :  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
6, Auxiliary tank (50 L) in PE-HD for demineralised water with automatic level control for external water feeding; compatible with demi water booster pump PAD/PAD2; water intake from auxiliary system
Dimensions (Ø×H): 340×640 mm, −
20 240,00 Kč
1 * 1 KS
  • Popis:
    Auxiliary tank (50 L) in PE-HD for demineralised water with automatic level control for external water feeding; compatible with demi water booster pump PAD/PAD2; water intake from auxiliary system
    Dimensions (Ø×H): 340×640 mm
  • Sekvence:
    6
  • Určeno pro: