Accessories for glassware machine washer
72, T6040, frame, open, stainless steel. Raises working level to 70 cm above floor level, W×D×H: 600×530×400 mm, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060P, GW3060S, GW4060, GW4060C, GW4060S, GW4060SC, 600 × 530 × 400 mm
Catalog # 141-0797
Supplier: SMEG
Jump to:
Product Details & Documents
Specifications
- Popis:T6040, frame, open, stainless steel. Raises working level to 70 cm above floor level, W×D×H: 600×530×400 mm
- Sekvence:72
- Š×H×V:600 × 530 × 400 mm
- Určeno pro:GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060P, GW3060S, GW4060, GW4060C, GW4060S, GW4060SC