Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
72, T6040, frame, open, stainless steel. Raises working level to 70 cm above floor level, W×D×H: 600×530×400 mm, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060P, GW3060S, GW4060, GW4060C, GW4060S, GW4060SC, 600 × 530 × 400 mm
Catalog # 141-0797
Supplier:   SMEG
Accessories for glassware machine washer
Catalog # 141-0797
Supplier:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
72, T6040, frame, open, stainless steel. Raises working level to 70 cm above floor level, W×D×H: 600×530×400 mm, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060P, GW3060S, GW4060, GW4060C, GW4060S, GW4060SC, 600 × 530 × 400 mm
Price
32 720,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    T6040, frame, open, stainless steel. Raises working level to 70 cm above floor level, W×D×H: 600×530×400 mm
  • Sekvence:
    72
  • Š×H×V:
    600 × 530 × 400 mm
  • Určeno pro:
    GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060P, GW3060S, GW4060, GW4060C, GW4060S, GW4060SC
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.