Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
Accessories for glassware machine washer
44, LBT5DS jet rack, stainless steel, with 5 nozzles and drying system, for 5 l bottles, max. 180×500 mm (Ø×H), GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Katalogové číslo 141-0729
Dodavatel:   SMEG
Accessories for glassware machine washer
Katalogové číslo 141-0729
Dodavatel:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
44, LBT5DS jet rack, stainless steel, with 5 nozzles and drying system, for 5 l bottles, max. 180×500 mm (Ø×H), GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Přejít na:
Price
68 470,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    LBT5DS jet rack, stainless steel, with 5 nozzles and drying system, for 5 l bottles, max. 180×500 mm (Ø×H)
  • Sekvence:
    44
  • Určeno pro:
    GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.