Accessories for glassware machine washer
44, LBT5DS jet rack, stainless steel, with 5 nozzles and drying system, for 5 l bottles, max. 180×500 mm (Ø×H), GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC
Katalogové číslo 141-0729
Dodavatel: SMEG
Přejít na:
Product Details & Documents
Specifikace
- Popis:LBT5DS jet rack, stainless steel, with 5 nozzles and drying system, for 5 l bottles, max. 180×500 mm (Ø×H)
- Sekvence:44
- Určeno pro:GW4060, GW4060C, GW4060S, GW4060SC, GW4090, GW4090C, GW4190, GW4190C, GW4190S, GW4190SC