Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
Accessories for glassware machine washer
95, Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, GW1160, GW1160C, GW1160S, GW3060BX, GW3060BXC,
GW3060P, GW3060S, GW3060SC
Katalogové číslo 141-0730
Dodavatel:   SMEG
Accessories for glassware machine washer
Katalogové číslo 141-0730
Dodavatel:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
95, Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, GW1160, GW1160C, GW1160S, GW3060BX, GW3060BXC,
GW3060P, GW3060S, GW3060SC
Přejít na:
Price
44 390,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares
  • Sekvence:
    95
  • Určeno pro:
    GW1160, GW1160C, GW1160S, GW3060BX, GW3060BXC,
    GW3060P, GW3060S, GW3060SC
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.