Accessories for glassware machine washer
95, Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, GW1160, GW1160C, GW1160S, GW3060BX, GW3060BXC,
GW3060P, GW3060S, GW3060SC
GW3060P, GW3060S, GW3060SC
Katalogové číslo 141-0730
Dodavatel: SMEG
Přejít na:
Product Details & Documents
Specifikace
- Popis:Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares
- Sekvence:95
- Určeno pro:GW1160, GW1160C, GW1160S, GW3060BX, GW3060BXC,
GW3060P, GW3060S, GW3060SC