Accessories for glassware machine washer
96, Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, with drying system, GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C
GW4090C
141-0731
: SMEG
- Popis:Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, with drying system
- Sekvence:96
- Určeno pro:GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C