Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
96, Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, with drying system, GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C
  141-0731
 :   SMEG
Accessories for glassware machine washer
  141-0731
 :  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
96, Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, with drying system, GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
GW4090C
49 590,00 Kč
1 * 1 KS
  • Popis:
    Upper universal flask washing, stainless steel, with 20 positions, for washing narrow necked glasswares, with drying system
  • Sekvence:
    96
  • Určeno pro:
    GW4060, GW4060C, GW4060S, GW4060SC, GW4090,
    GW4090C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.