Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Accessories for glassware machine washer
45, LM40 universal jet rack, stainless steel, 40 assorted nozzles, for washing narrow necked glassware, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060,P GW3060S
Katalogové číslo 141-0734
Dodavatel:   SMEG
Accessories for glassware machine washer
Katalogové číslo 141-0734
Dodavatel:  SMEG
Accessories for glassware machine washer
Accessories for glassware machine washer
45, LM40 universal jet rack, stainless steel, 40 assorted nozzles, for washing narrow necked glassware, GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060,P GW3060S
Přejít na:
Price
48 240,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    LM40 universal jet rack, stainless steel, 40 assorted nozzles, for washing narrow necked glassware
  • Sekvence:
    45
  • Určeno pro:
    GW0160, GW0160S, GW1160, GW1160C, GW1160S, GW1160SC, GW3060BX, GW3060BXC, GW3060,P GW3060S
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.