Power selector, WRT00116
01, Power selector, WRT00116
Katalogové číslo WINKWRT00116-XX013
Dodavatel: WINKLER
Přejít na:
Product Details & Documents
Plug-in power selector made of PS with switching mechanism.
- Rotary switch with bimetal element
- Signal light for controlling status indication
- Cable with grounding plug type
Specifikace
- Model:WRT00116
- Popis:Power selector
- Sekvence:01
- Cat. No.:WINKWRT00116-XX013
- Š×H×V:120×56×57 mm
- Elektrické napětí:230 V; 13 A (ohmic load); 50 Hz