Pipette tips, ClickSure™ Tips
01, Standard, 50 µl, -, Bulk, 960/bag
Katalogové číslo 613-2933
Dodavatel: HAMILTON BONADUZ
Přejít na:
Product Details & Documents
These tips are validated to optimise the performance of the Microlab 300 system, offering superior accuracy and precision. When the tips are placed onto the probe, the O-ring on the probe compresses into the tip groove. This design only requires a low insertion force to successfully load a tip, resulting in a more ergonomic experience for the user and enabling long-term use with minimal energy exerted.
- Tip packaging and tips are recyclable for eco-conscious labs
- Less force is required to load and eject a tip
Specifikace
- Balené:Bulk, 960/bag
- Sterilní:-
- Objem:50 µl
- Sekvence:01
- Druh špičky:Standard