Mops, LoopMop™
LoopMop™, irradiated
Katalogové číslo MICRMZLM5-12IR
Dodavatel: MICRONOVA
Přejít na:
Product Details & Documents
Knitted polyester mop for applying disinfectant in pharmaceutical, aseptic processing and medical device applications. Absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. Mop heads can be used directly with the fibreglass/polypropylene handle or attached to the universal handle via the quick disconnect adapter.
- Can be laundered or autoclaved
- Low linting
- No raw edges
Informace pro objednávání: All components must be ordered separately.
Specifikace
- Popis:LoopMop™, irradiated
- Cat. No.:MICRMZLM5-12IR
- Environmentally Preferable:Waste Reducing
Accessories
Informace o produktu
- Katalogové čísloSekvenceDélkaDostupnostPrice
- Prodlužovací nerezová tyčProdlužovací nerezová tyč129-055101965 - 1829 mmPřihlásit se pro dostupnost1 * 1 KS - 7 840,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDélkaDostupnostPrice
- Standardní nerezová tyč
Informace o produktu
- Katalogové čísloSekvenceDélkaDostupnostPrice
- Stainless steel quick disconnect handleStainless steel quick disconnect handle129-0176081295 cmPřihlásit se pro dostupnost1 * 1 KS - 3 490,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesMicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles129-017004Přihlásit se pro dostupnost1 * 1 KS - 3 430,00 Kč