TEST SUBSTANCE SACCHARIN 1 * 5 g
Catalog # METN51143091
Supplier: Mettler - Toledo
Frequently Bought Together
Pyridin, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyklohexanon, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Glycerin ≥97%, TECHNICAL
Supplier: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Diethylenglykolmonobutylether, Sigma-Aldrich®
Supplier: Merck
Voda p.a.
Supplier: Thermo Fisher Scientific
Dichlormethan ≥99.7% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Mineral oil light USP, NF, BP, Ph. Eur., pharma grade
Supplier: PanReac AppliChem
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Butanon 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific