Orders LinkContactUsLinkComponent
Roll mop, PolyTack®
Roll mop, PolyTack®
03, Tacky roller, foam for contoured surfaces, 457,2 mm, 20 sheets/roll
Katalogové číslo CONNPR-18F
Dodavatel:   CONNECTICUT CLEANROOM
Roll mop, PolyTack®
Katalogové číslo CONNPR-18F
Roll mop, PolyTack®
Roll mop, PolyTack®
03, Tacky roller, foam for contoured surfaces, 457,2 mm, 20 sheets/roll
Přejít na:
Price
2 280,00 Kč
Přihlásit se pro dostupnost
1 * 1 Roll
Product Details & Documents
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
  • Easy to use hand or pole attachments
  • Plastic inner core insures cleanliness
  • Anti-microbial agent providing protection against bacterial growth
Specifikace
  • Popis:
    Tacky roller, foam for contoured surfaces, 457,2 mm
  • Sekvence:
    03
  • Balené:
    20 sheets/roll