Orders LinkContactUsLinkComponent
Roll mop, PolyTack®
Roll mop, PolyTack®
05, Roller mop handle, stainless steel, for 457,2 mm mops, -
Catalog # CONNPH-18
Roll mop, PolyTack®
Catalog # CONNPH-18
Roll mop, PolyTack®
Roll mop, PolyTack®
05, Roller mop handle, stainless steel, for 457,2 mm mops, -
Price
8 990,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
  • Easy to use hand or pole attachments
  • Plastic inner core insures cleanliness
  • Anti-microbial agent providing protection against bacterial growth
Specifications
  • Popis:
    Roller mop handle, stainless steel, for 457,2 mm mops
  • Sekvence:
    05
  • Balené:
    -