Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
08, with polypropylene screw cap, individually wrapped, labelled, irradiated, 150 ml
Katalogové číslo 216-2052
Dodavatel:   Anicrin
Jars with screw cap
Katalogové číslo 216-2052
Dodavatel:  Anicrin
Jars with screw cap
Jars with screw cap
08, with polypropylene screw cap, individually wrapped, labelled, irradiated, 150 ml
Přejít na:
Price
2 760,00 Kč
Přihlásit se pro dostupnost
1 * 250 KS
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Specifikace
  • Type:
    with polypropylene screw cap, individually wrapped, labelled, irradiated
  • Kapacita:
    150 ml
  • Sekvence:
    08