Orders LinkContactUsLinkComponent
Dissection boards, disposable
Dissection boards, disposable
02, Disposable dissection boards, Bílá, 333×233 mm
Katalogové číslo LPIT194032
Dodavatel:   LP ITALIANA
Dissection boards, disposable
Katalogové číslo LPIT194032
Dodavatel:  LP ITALIANA
Dissection boards, disposable
Dissection boards, disposable
02, Disposable dissection boards, Bílá, 333×233 mm
Přejít na:
Price
12 040,00 Kč
Přihlásit se pro dostupnost
1 * 48 KS
Product Details & Documents
Disposable cutting boards made of cardboard and PS foam for the dissection of surgical specimens.
  • With data registration area
  • Pre-printed scale (in mm)
  • Three different sizes available
Specifikace
  • Barva:
    Bílá
  • Popis:
    Disposable dissection boards
  • D׊:
    333×233 mm
  • Sekvence:
    02