[EN]METALLIC PARTICULATE FILTER ACC 1 * 1 KS
Katalogové číslo BWTEGA-MFILTER
Dodavatel: BW TECHNOLOGIES
[EN]METALLIC PARTICULATE FILTER ACC 1 * 1 KS
Katalogové číslo BWTEGA-MFILTER
Dodavatel: BW TECHNOLOGIES
Frequently Bought Together
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
Hydroxid sodný 0.1 mol/l (0.1 N) vodný roztok, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Dodavatel: VWR Chemicals
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
VWR® NITRILE LIGHT, Rukavice, nitrilové
Dodavatel: VWR Collection