Orders LinkContactUsLinkComponent
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
36, Swaged post, stainless steel with aluminium cap, 1600 mm dimension, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
Catalog # METO63UPS-SW
Supplier:   Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Catalog # METO63UPS-SW
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
36, Swaged post, stainless steel with aluminium cap, 1600 mm dimension, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
Price
3 300,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Swaged post, stainless steel with aluminium cap, 1600 mm dimension
  • Sekvence:
    36
  • Určeno pro:
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving