REGENERATION SOLUTION 1 * 25 mL
Catalog # METN51350104
Supplier: Mettler - Toledo
Frequently Bought Together
Cyklohexan, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Voda, GPR RECTAPUR®
Supplier: VWR Chemicals
Voda s 0,1% (v/v) Kyselina mravenčí, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
n-Heptan ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
terc-Butylmethylether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
jod 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Butanon, EMSURE® ACS, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Toluen ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dichlormethan ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dimethylsulfoxid, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Pyridin, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,4-Dioxan, extra dry over Molecular Sieve 99,5% stabilizovaný, AcroSeal®
Supplier: Thermo Fisher Scientific