Salicylan sodný p.a., (Xn) 1 * 500 g
Katalogové číslo LACH221430778050
Dodavatel: Lach-ner S.R.O.
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Jodid draselný ≥99.5%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Chlorid draselný, EMSURE® p.a., Supelco®
Dodavatel: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Dihydrogenfosforečnan draselný, EMSURE® ISO p.a., Supelco®
Dodavatel: Merck
Hydrogenfosforečnan didraselný, EMSURE® p.a., Supelco®
Dodavatel: Merck
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Chlorid sodný regeneration salt, TECHNICAL for water softening
Dodavatel: VWR Chemicals
Chlorid sodný 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Kyselina sírová 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Chlornan sodný 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Uhličitan sodný, anhydrous, EMSURE® ISO p.a., Supelco®
Dodavatel: Merck
pH buffer solutions, colour coded
Dodavatel: REAGECON