Erlenmeyer Flasks, Narrow Neck
08, 2000 ml, 280 mm, 166 mm, 50 mm
GLAS231.202.07
: GLASSCO
Clear, conical, graduated and narrow mouth Erlenmeyer flask, made from ASTM E-438 Type 1, Borosilicate 3.3 heat resistance glass.
- All the flasks are printed in white enamel
- Has reinforced thick rim to reduce chipping and breakage
: Complies with ISO 1773.
- Ø vněj.:166 mm
- Kapacita:2000 ml
- Výška:280 mm
- Sekvence:08
- Hrdlo rozš. Ø:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



