Tanks, rectangular, LDPE, LaboPlast®
25, Lid, LDPE, for rectangular tanks, 90 L
Katalogové číslo BURK3440-0901
Dodavatel: BURKLE
Přejít na:
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
- Linear LDPE, transparent
- Rupture-proof
Specifikace
- Popis:Lid, LDPE, for rectangular tanks, 90 L
- Sekvence:25
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Container thread ¾" for cylindrical containersContainer thread ¾" for cylindrical containersBURK3440-003401Přihlásit se pro dostupnost1 * 1 KS - 4 360,00 Kč