Tanks, rectangular, LDPE, LaboPlast®
03, 45 L, Seamless containers, rectangular shaped, - mm
Katalogové číslo BURK3440-0455
Dodavatel: BURKLE
Přejít na:
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
- Linear LDPE, transparent
- Rupture-proof
Specifikace
- Kapacita:45 L
- Popis:Seamless containers, rectangular shaped
- Sekvence:03
- Š×H×V:- mm
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Container thread ¾" for cylindrical containersContainer thread ¾" for cylindrical containersBURK3440-003401Přihlásit se pro dostupnost1 * 1 KS - 4 360,00 Kč