Tween® 80 (Polysorbate)
3745, Plastová láhev, 1.000 g
Catalog # LACH40164BT0G1000
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:13,07664
- Boiling Pt:∼100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1,062 g/cm³ (20 °C)
- Flash Pt:>149 °C
- :Density: 1,062 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-019-9
Specifications
Specifications
- Typ balení:Plastová láhev
- Bal.:1.000 g
- Sekvence:3745



