Murexide
3236, Glass bottle for solids, 10 g
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19
- Merck Index:13,06330
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Documents
Specifications
Specifications
- Typ balení:Glass bottle for solids
- Bal.:10 g
- Sekvence:3236
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



