Tween® 80 (Polysorbate)
3645, Plastová láhev, 500 g
LACH40164FT0G0500
- :9005-65-6
- :MFCD00082107
- :13,07664
- :∼100 °C (1013 hPa)
- :–21 °C
- :1,062 g/cm³ (20 °C)
- :>149 °C
- :Density: 1,062 g/cm³ (20 °C)
- :Ambient
- :500-019-9
- Typ balení:Plastová láhev
- Bal.:500 g
- Sekvence:3645
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.