Chlorid hydroxylamonný
3545, Plastová láhev, 250 g
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49
- Merck Index:13,04853
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:151 °C
- Density:1,67 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:226-798-2
- ADR:8,III
- :Density: 1,67 g/cm³ (20 °C)
- UN:2923
Documents
Specifications
Specifications
- Typ balení:Plastová láhev
- Bal.:250 g
- Sekvence:3545



