Hydrogendifluorid amonný
3745, Plastová láhev, 1.000 g
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Ammonium bifluoride, Ammonium hydrogen fluoride
- CAS Number:1341-49-7
- Formula:NH₄HF₂
- MDL Number:MFCD00012018
- Molecular Weight:57.04
- Merck Index:13,00498
- Melting Pt:127 °C
- Density:0,8 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-676-4
- UN:1727
- ADR:8,II
- :Density: 0,8 g/cm³ (20 °C)
Documents
Specifications
Specifications
- Typ balení:Plastová láhev
- Bal.:1.000 g
- Sekvence:3745



