Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
n-Hexan

n-Hexan

5037, Glass bottle, 1 L
Catalog # LACH20031CT0M1000
Flammable
Health
Irritant
Environment
Supplier:   Lach-ner S.R.O.
n-Hexan
Catalog # LACH20031CT0M1000
Supplier:  Lach-ner S.R.O.
CAS Number:  110-54-3
n-Hexan
n-Hexan

5037, Glass bottle, 1 L

CAS Number:  110-54-3
Price
295,00 Kč
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • UN:1208
  • ADR:3,II
  •  :Density: 0,659 g/cm³ (20 °C)
  • EINECS:203-777-6
Specifications

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    1 L
  • Sekvence:
    5037

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.