Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Síran amonný

Síran amonný

3645, Plastová láhev, 500 g
Catalog # LACH30165AP0G0500
Supplier:   Lach-ner S.R.O.
Síran amonný
Catalog # LACH30165AP0G0500
Supplier:  Lach-ner S.R.O.
CAS Number:  7783-20-2
Síran amonný

3645, Plastová láhev, 500 g

CAS Number:  7783-20-2
Price
155,00 Kč
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
  • CAS Number:7783-20-2
  • Formula:(NH₄)₂SO₄
  • MDL Number:MFCD00003391
  • Molecular Weight:132.14
  • Merck Index:13,00559
  • Melting Pt:336…339 °C
  • Density:1,78 g/cm³ (20 °C)
  •  :Density: 1,78 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-984-1
  • REACH:01-2119455044-46
Specifications

Specifications

  • Typ balení:
    Plastová láhev
  • Bal.:
    500 g
  • Sekvence:
    3645

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.