Dichlormethan
5037, Glass bottle, 1 L
LACH20020CT1M1000
: Methylene chloride, DCM
- :75-09-2
- :CH₂Cl₂
- :MFCD00000881
- :84.93
- :12,06140
- :39,8 °C (1013 hPa)
- :–95 °C
- :1,322 g/cm³ (20 °C)
- :Ambient
- :200-838-9
- :6.1,III
- :Density: 1,322 g/cm³ (20 °C)
- :1593
- Typ balení:Glass bottle
- Bal.:1 L
- Sekvence:5037
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.