Síran stříbrný
3401, 100 g
: MACH
: Disilver sulphate
- :10294-26-5
- :Ag₂SO₄
- :MFCD00003407
- :311.8
- :13,08603
- :1085 °C (1013 hPa)
- :655 °C
- :5,45 g/cm³ (20 °C)
- :Density: 5,45 g/cm³ (20 °C)
- :Ambient
- :233-653-7
- Bal.:100 g
- Sekvence:3401
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.