Vinan antimonylo-draseln hemihydrt p.a., 100 g (Xn,N) 1 * 100 g
Katalogové číslo MACH121-949-0100
Dodavatel: MACH
Vinan antimonylo-draseln hemihydrt p.a., 100 g (Xn,N) 1 * 100 g
Katalogové číslo MACH121-949-0100
Dodavatel: MACH
Frequently Bought Together
Salicylát sodný ≥99.5%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Chlorid sodný 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Chlornan sodný 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals