Kyselina chlorovodíková N/2, c=0,5mol/l ORS, 1000 ml (C) 1 * 1 L
Catalog # MACH121-019-1000
Supplier: MACH
Kyselina chlorovodíková N/2, c=0,5mol/l ORS, 1000 ml (C) 1 * 1 L
Catalog # MACH121-019-1000
Supplier: MACH
Frequently Bought Together
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Ubrousky čisticí
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Rukavice, nitrilové
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection