ROZTOK PRO ZELENYHO 1 * 1 L
Katalogové číslo MACH121011-1000
Dodavatel: MACH
Frequently Bought Together
Glycerin ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Dodavatel: VWR Chemicals
Heptan, TECHNICAL
Dodavatel: VWR Chemicals
Dimethylsulfoxid ≥99.0%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Toluen ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Diethylether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Ethanol eurodenatured ≥80%, TechniSolv® (1:1:1)
Dodavatel: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Dodavatel: VWR Chemicals
Diisodecyl phthalate, Sigma-Aldrich®
Dodavatel: Merck
Butanon ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Voda ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Dodavatel: VWR Chemicals
Dimethylsulfoxid, EMSURE® ACS p.a., Supelco®
Dodavatel: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Dodavatel: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Chloroform 99+% stabilizovaný, extra pure
Dodavatel: Thermo Fisher Scientific
Boron trifluoride - methanol complex 10% in methanol, Supelco®
Dodavatel: Merck
Glycerin, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Dodavatel: Honeywell Chemicals