Orders LinkContactUsLinkComponent
Biological indicators, spore threads
Biological indicators, spore threads
Spore threads, bulk packed
Katalogové číslo NAMSTHS-06
Dodavatel:   CROSSTEX
Biological indicators, spore threads
Katalogové číslo NAMSTHS-06
Dodavatel:  CROSSTEX
Biological indicators, spore threads
Spore threads, bulk packed
Přejít na:
Price
31 520,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
Spore threads are prepared from Geobacillus stearothermophilus. These are designed to be placed directly into a device and utilised to monitor steam sterilisation process efficacy.
Ideal for use in tubing, lumens and other devices/instruments where larger BIs and carriers cannot be utilised.
Specifikace
  • Popis:
    Spore threads, bulk packed