Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
500 ml, 19/26, 270 mm, 0,25 ± ml
Katalogové číslo VITL675950
Dodavatel:   VITLAB
Volumetric flasks, class A
Katalogové číslo VITL675950
Dodavatel:  VITLAB
Volumetric flasks, class A
Volumetric flasks, class A
500 ml, 19/26, 270 mm, 0,25 ± ml
Přejít na:
Price
2 000,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
PMP, opaque, with NS stoppers, PP.
  • With ring mark individually calibrated to ‘In’
  • With printed lot number and batch certificate
  • Thermal stress up to 121 °C (autoclaving) does not permanently exceed the tolerance limit
  • To preserve markings, cleaning at no higher than 60 °C is recommended
UV-absorbing, highly transparent. For storage of light-sensitive substances.
Certifications: Class A tolerances according to DIN EN ISO 1042.
Specifikace
  • NZ:
    19/26
  • Kapacita:
    500 ml
  • Výška:
    270 mm
  • Tolerance:
    0,25 ± ml