Orders LinkContactUsLinkComponent
Conductivity standards
01, Sodium chloride standard solution, 49 mg/l NaCl, 100 µS/cm, 500 mL
  HACH2971849
 :   Hach
Conductivity standards
  HACH2971849
 :  Hach
 :  
Conductivity standards
Conductivity standards
01, Sodium chloride standard solution, 49 mg/l NaCl, 100 µS/cm, 500 mL
 :  
1 620,00 Kč
1 * 500 mL
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
  • Bal.:
    500 mL
  • Popis:
    Sodium chloride standard solution, 49 mg/l NaCl
  • Sekvence:
    01
  • Vodivost:
    100 µS/cm