Buffers, sodium phosphate, 0,2 M solutions
02, Sodium phosphate buffer solution, 0,2 mol/l, 7,0, Glass bottle, 1 L
Physical form: liquid
MDL No.: MFCD00146206
- Koncentrace:0,2 mol/l
- Typ balení:Glass bottle
- Bal.:1 L
- Popis:Sodium phosphate buffer solution
- Sekvence:02
- Hodnota pH:7,0
- Cat. No.:J63482.K2
- Fyzická forma:Liquid
- MDL čí.:MFCD00146206
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



