Reagent 3 rozpuštěný kyslík, práškový 1 * 25 KS
Katalogové číslo HACH98768
Dodavatel: Hach
Frequently Bought Together
Diethylether, EMPARTA® ACS p.a., Supelco®
Dodavatel: Merck
Pyridin ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Dichlormethan ≥99% stabilised, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Diethylenglykolmonobutylether, Sigma-Aldrich®
Dodavatel: Merck
Ethylacetát ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochlorid, Supelco®
Dodavatel: Merck
N,N-Dimethylformamid 99+%, extra pure
Dodavatel: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Dodavatel: Thermo Fisher Scientific
jodmetan ≥99% stabilizovaný
Dodavatel: Thermo Fisher Scientific
1,4-Dioxan, extra dry over Molecular Sieve 99,5% stabilizovaný, AcroSeal®
Dodavatel: Thermo Fisher Scientific
N,N-Dimethylformamid, extra dry over Molecular Sieve 99.8%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
n-Oktan, extra dry 99+%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Aceton ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Dodavatel: PanReac AppliChem
n-Oktan ≥98%
Dodavatel: Thermo Fisher Scientific
Cyklohexan ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals