Orders LinkContactUsLinkComponent
Labels, biohazard
Labels, biohazard
2, Biohazard labels, Polyester, Černá na žluté, 100×87 mm, 3 labels/card
Catalog # 116-0751
Supplier:   Brady
Labels, biohazard
Catalog # 116-0751
Supplier:  Brady
Labels, biohazard
Labels, biohazard
2, Biohazard labels, Polyester, Černá na žluté, 100×87 mm, 3 labels/card
Price
319,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
All signs comply with current European legislation.
Ordering information: The polyester labels have a permanent acrylic adhesive.
Specifications
  • Barva:
    Černá na žluté
  • Popis:
    Biohazard labels
  • Rozměry:
    100×87 mm
  • Materiál:
    Polyester
  • Sekvence:
    2
  • Balené:
    3 labels/card