Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
24/40, 65 mm
Catalog # COWI010.1124
Supplier:   COWIE
Stoppers, self realasing, PTFE
Catalog # COWI010.1124
Supplier:  COWIE
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
24/40, 65 mm
Price
893,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specifications
  • NZ:
    24/40
  • Výška:
    65 mm