Orders LinkContactUsLinkComponent
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
06, Stationary post, SiteSelect™, stainless steel, 875 mm height, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
Katalogové číslo 139-0257
Dodavatel:   Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Katalogové číslo 139-0257
Dodavatel:  Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
06, Stationary post, SiteSelect™, stainless steel, 875 mm height, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
Přejít na:
Price
2 140,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Stationary post, SiteSelect™, stainless steel, 875 mm height
  • Sekvence:
    06
  • Určeno pro:
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving