Orders LinkContactUsLinkComponent
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
10, Mobile post, SiteSelect™, stainless steel, 857 mm height, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
Catalog # 139-0273
Supplier:   Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Catalog # 139-0273
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
10, Mobile post, SiteSelect™, stainless steel, 857 mm height, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
Price
2 430,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Mobile post, SiteSelect™, stainless steel, 857 mm height
  • Sekvence:
    10
  • Určeno pro:
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving