Orders LinkContactUsLinkComponent
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
16, Mobile post, SiteSelect™, stainless steel, 1385 mm height, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
Katalogové číslo 139-0274
Dodavatel:   Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Katalogové číslo 139-0274
Dodavatel:  Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
16, Mobile post, SiteSelect™, stainless steel, 1385 mm height, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
Přejít na:
Price
2 640,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Mobile post, SiteSelect™, stainless steel, 1385 mm height
  • Sekvence:
    16
  • Určeno pro:
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving