Orders LinkContactUsLinkComponent
Sample containers, with screw cap, universal, WHEATON®
Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, pre-filled with boric acid for urine samples, printed label, sterile, Průsvitná, PP/PE
Katalogové číslo 216-0989
Dodavatel:   DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Katalogové číslo 216-0989
Dodavatel:  DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, pre-filled with boric acid for urine samples, printed label, sterile, Průsvitná, PP/PE
Přejít na:
Price
3 690,00 Kč
Přihlásit se pro dostupnost
1 * 400 KS
Product Details & Documents
PP translucent or PS clear, with PE screw cap.
  • Manufactured from FDA approved virgin polystyrene or polypropylene
  • PS offers good thermal and chemical resistance
  • Supplied sterile - for in vitro use only
  • All materials are non cytotoxic
Straight walled containers ideal for sample collection, storage and transport. Supplied with leakproof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certifications: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Balení: Packed in inner bags of 50.
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Specifikace
  • Type:
    Universal container, pre-filled with boric acid for urine samples, printed label, sterile
  • Kapacita:
    30 ml
  • Barva:
    Průsvitná
  • Materiál:
    PP/PE