Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS
Katalogové číslo LENZ04018037
Dodavatel:   Lenz Laborglas
Separating funnels, Squibb
Katalogové číslo LENZ04018037
Dodavatel:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS
Přejít na:
Price
4 810,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids
Documents
Specifikace
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml