Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
n-Hexan ≥98%, SupraSolv® for GC-MS

n-Hexan ≥98%, SupraSolv® for GC-MS

5037, Glass bottle, 1 L
Catalog # 1.00795.1000
Flammable
Health
Irritant
Environment
Supplier:   Merck
n-Hexan ≥98%, SupraSolv® for GC-MS
Catalog # 1.00795.1000
Supplier:  Merck
CAS Number:  110-54-3
n-Hexan ≥98%, SupraSolv® for GC-MS
n-Hexan ≥98%, SupraSolv® for GC-MS

5037, Glass bottle, 1 L

CAS Number:  110-54-3
Price
1 230,00 Kč
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 0,659 g/cm³ (20 °C)
Specifications

Specification Test Results

  • Purity (GC) (sum of hexane isomers plus methylcyclopentane):
    =98.0% (=99.8%)
  • Identity(IR):
    Conforms
  • Evaporation Residue:
    = 3.0 mg/l
  • Water:
    = 0.01%
  • Color:
    =10 Hazen
  • GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):
    = 3ng/ml
  • Suitability:
    Residue Analysis

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    1 L
  • Sekvence:
    5037

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.