Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
n-Hexan ≥98%, SupraSolv® for GC-MS

n-Hexan ≥98%, SupraSolv® for GC-MS

5096, Glass bottle, 2,5 L
Catalog # 1.00795.2500
Flammable
Health
Irritant
Environment
Supplier:   Merck
n-Hexan ≥98%, SupraSolv® for GC-MS
Catalog # 1.00795.2500
Supplier:  Merck
CAS Number:  110-54-3
n-Hexan ≥98%, SupraSolv® for GC-MS
n-Hexan ≥98%, SupraSolv® for GC-MS

5096, Glass bottle, 2,5 L

CAS Number:  110-54-3
Price
2 590,00 Kč
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 0,659 g/cm³ (20 °C)
Specifications

Specification Test Results

  • Purity (GC) (sum of hexane isomers plus methylcyclopentane):
    =98.0% (=99.8%)
  • Identity(IR):
    Conforms
  • Evaporation Residue:
    = 3.0 mg/l
  • Water:
    = 0.01%
  • Color:
    =10 Hazen
  • GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)):
    = 3ng/ml
  • Suitability:
    Residue Analysis

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    2,5 L
  • Sekvence:
    5096

Specifications